Kimble™ Glass Dropping Bottles
Actual product may vary
DWK Life Sciences
Kimble™ Glass Dropping Bottles
Convenient all-in-one configuration
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-992B60 mL
FIS02-992A30 mL
Catalog number FIS02-992B
Price (SGD)
-
Capacity (Metric):
60 mL
Convenient all-in-one configuration
  • Constructed of GG-33 borosilicate glass
  • Ground-in glass pipet and rubber bulb included
Specifications
Capacity2.02 oz., 60mL
Capacity (Metric)60 mL
Capacity (English)2.02 oz.
IncludesGround in Glass pipet and rubber bulb