Amber Wide Mouth Packer Bottles with Green Thermoset F217 and PTFE Cap
Actual product may vary
Qorpak
Amber Wide Mouth Packer Bottles with Green Thermoset F217 and PTFE Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Glass Jug with Green Thermoset F217 and PTFE Cap are used in chemical and environmental applications.
Have Questions?
Catalog NumberCapacity (Metric)Quantity
FIS02-992-63115 mL48/Cs.
Catalog number FIS02-992-631
Price (SGD)
342.00
Case
Capacity (Metric):
15 mL
Quantity:
48/Cs.
Price (SGD)
342.00
Case
  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • Classic 'vitamin bottles' and are often used in pharmaceutical packaging
  • Closure shows high temperature tolerance with wide range of chemical compatibility
Specifications
Capacity0.5 oz., 15 mL
Capacity (Metric)15 mL
Capacity (English)0.5 oz.
ColorAmber
Diameter (English)1.22 in.
MaterialGlass
Quantity48/Cs.
SterilityNon-sterile
Closure ColorGreen
Closure IncludedYes
GraduatedNo
MouthWide, Threaded
ShapeRound
Height (Metric)50 mm
Product TypeWide Mouth Packer Bottle
Height (English)1.97 in.
Closure LinerF217, PTFE
Closure MaterialThermoset
Closure Size28-400 mm
Diameter (Metric)31 mm
Closure TypeScrew Cap