Check Order Status
Kimble™ Amber Dropping Bottles
Actual product may vary
DWK Life Sciences
Kimble™ Amber Dropping Bottles
With 1mL dropper
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Includes
FIS02-991D
60 mL
Plastic Dropper
FIS02-991A
30 mL
Glass Dropper
FIS02-991C
60 mL
Glass Dropper
3 Options
Catalog number
FIS02-991D
Price (SGD)
165.00
CS
Add to cart
Capacity (Metric):
60 mL
Includes:
Plastic Dropper
Price (SGD)
165.00
CS
Add to cart
With 1mL dropper
ASTM No. 438, Type II
Specifications
Capacity
2.02 oz., 60 mL
Capacity (Metric)
60 mL
Capacity (English)
2.02 oz.
Color
Amber
Diameter (English)
1.53 in.
Includes
Plastic Dropper
Material
Borosilicate Glass
Dropper Capacity
1 mL
Height (Metric)
118 mm
Product Type
Amber Dropping Bottle
Height (English)
4.64 in.
Closure Size
20-400 mm
Diameter (Metric)
39 mm
Closure Type
Screw Cap