PYREX™ Aspirator Bottles with Tubulation
Actual product may vary
PYREX™ Aspirator Bottles with Tubulation
Capacity Metric: 9500 mL
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-972D1 L
Catalog number FIS02-972D
Price (SGD)
331.00
Each
Capacity (Metric):
1 L
Price (SGD)
331.00
Each
A range of bottles featuring a tubular sidearm outlet to facilitate attachment of flexible tubing. Useful as a delivery/storage container for solutions.
Specifications
Capacity (Metric)1 L
Capacity (English)33.81 oz.
Diameter (English)4.33 in.
Diameter (English) Outer Neck1.77 in.
Diameter (Metric) Outer Neck45 mm
For Use With (Application)Designed to store and discharge liquids
IncludesTubular sidearm outlet
MaterialBorosilicate Glass
No. per Case6/Cs.
No. per Pack1/Pk.
Stopper Number6
DisposableReusable
MouthWide
ShapeCylindrical
Height (Metric)200 mm
Product TypeAspirator Bottle
Height (English)7.87 in.
Closure MaterialPolypropylene
Diameter (Metric)110 mm
Closure TypeScrew Cap
Diameter (English) Outer Tubing0.39 in.
Diameter (Metric) Outer Tubing10 mm