PYREX™ Reagent Bottles with Hollow standard taper Stoppers
PYREX™ Reagent Bottles with Hollow standard taper Stoppers
Actual product may vary
PYREX™ Reagent Bottles with Hollow standard taper Stoppers
For storing reagents, sera, vaccines, colloidal solutions and similar liquids because of their outstanding chemical durability
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Stopper Number
FIS02-940C250 mL19
FIS02-940B125 mL19
Catalog number FIS02-940C
Price (SGD)
115.00
EA
Add to cart
Capacity (Metric):
250 mL
Stopper Number:
19
Price (SGD)
115.00
EA
Add to cart
With neck ground and furnished with standard taper stopper.
For stopper only, see Cat. No. 7575.
Specifications
Capacity8.45 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
Diameter (English)2.75 in.
Diameter (English) Outer Neck1.22 in.
Diameter (Metric) Outer Neck31 mm
MaterialBorosilicate Glass
No. per Case36/Cs.
No. per Pack1/Pk.
SterilityNon-sterile
Stopper Number19
Closure ColorOrange
MouthNarrow
ShapeSquare
Height (Metric)160 mm
Height (English)6.29 in.
Closure MaterialPolypropylene
Diameter (Metric)70 mm
Closure TypePlug Seal
Unit SizeEA