Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Size |
|---|---|---|
| FIS02-925-3A | 30 mL | 28 mm |
| 2106-0001 | 30 mL | 28 mm |
| 2106-0002 | 60 mL | 28 mm |
| FIS02-925-3B | 60 mL | 28 mm |
| FKS10547692 | 60 mL | 28 mm |
| 2106-0004 | 125 mL | 38 mm |
| FIS02-925-3C | 125 mL | 38 mm |
| 2106-0008 | 250 mL | 43 mm |
| FIS02-925-3D | 250 mL | 43 mm |
| 2106-0016 | 500 mL | 53 mm |
| FIS02-924-22 | 500 mL | 53 mm |
| 2106-0032 | 1 L | 63 mm |
| FIS02-925-3E | 1 L | 63 mm |
Reduce UV light transmissions to protect light-sensitive contents with Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality Amber HDPE Bottles. Useful for storage, shipping and packaging, they provide excellent chemical resistance to most acids, bases and alcohols. These high-density polyethylene bottles feature an easy-to-fill wide mouth and are leakproof† when used with Nalgene closures.
Includes:
Amber polypropylene screw caps
.
Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.