Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure
Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure
Actual product may vary
Thermo Scientific
Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure
Get excellent resistance to temperature, chemicals and corrosion with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure, among the most chemical- and corrosion-resistant containers available.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS02-923-35L250 mL38-430 mm
FKS1591166160 mL20 mm
FKS15911381125 mL38-430 mm
FIS02-923-35N1 L38-430 mm
Catalog number FIS02-923-35L
Price (SGD)
2,495.00
CS
Add to cart
Capacity (Metric):
250 mL
Closure Size:
38-430 mm
Price (SGD)
2,495.00
CS
Add to cart
Get excellent resistance to temperature, chemicals and corrosion with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure, among the most chemical- and corrosion-resistant containers available.
  • Withstand -270°C to +250°C (-454°F to +482°F)
  • Inert to virtually all chemicals except molten alkali metals, fluorine at high temperatures and complex halogenated compounds at high temperatures and pressures
  • Packaged individually
  • Autoclavable
  • Leakproof† PFA closure
Includes:
Leakproof, linerless PFA screw caps
Note:
Completely disengage threads or remove cap before autoclaving
Warranty:
90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
DescriptionNarrow Mouth Bottle
Diameter (Metric) Inner Neck24 mm
For Use With (Application)Storage, Transportation ​
MaterialTeflon PFA
No. per Case4/Cs.
No. per Pack1 Ea.
SterilityNon-sterile
Temperature Range (Metric)-100°C to 260°C
Closure ColorNatural
Neck Finish38 mm
Certifications/ComplianceCCD 077, SW-CDC-001501, EN-8-1007-22-0912
Closure IncludedYes
AutoclavableAutoclavable
MouthNarrow
ShapeRound
Height (Metric)146 mm
Product TypeNarrow Mouth Bottle
Height (English)5.74 in.
Closure LinerUnlined
Closure MaterialETFE
Closure Size38-430 mm
Diameter (Metric)59 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS