Specifications
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorClear
For Use With (Application)Soil sampling and environmental applications
MaterialSoda Lime Glass
Quantity24/PK.
Closure ColorWhite
Certifications/ComplianceEPA
Closure IncludedYes
Closure LinerPTFE
Closure Size70-400 mm
Closure TypeScrew Cap
AutoclavableAutoclavable (120°C)
Unit SizeCase