Certified Low Particulate Wide Mouth Packer Bottles
Actual product may vary
Certified Low Particulate Wide Mouth Packer Bottles
Ideal for the storage of dry powders. Fisherbrand™ Certified Low Particulate Wide Mouth Packer Bottles are manufactued from clear soda glass.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FIS02-912-040250 mL58-400 mm
Catalog number FIS02-912-040
Price (SGD)
621.00
Pack
Capacity (Metric):
250 mL
Closure Size:
58-400 mm
Price (SGD)
621.00
Pack
Ideal for the storage of dry powders. Fisherbrand™ Certified Low Particulate Wide Mouth Packer Bottles are manufactued from clear soda glass.
  • Graduated clear glass
  • Wide mouth allows for easy accessibility
  • With white polypropylene cap with PTFE faced foamed polyethylene liner
  • Certified to be cleaned for low particulates, certificate of analysis attached
Specifications
Capacity8.45 oz., 250mL
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
Cleaning ProcedureUSP purified Water
ColorClear
MaterialGlass
Length (Metric)70mm
Quantity24/PK.
Certifications/ComplianceCertified low particulate
Closure IncludedYes
Product TypeGraduated Bottle
Closure MaterialPTFE
Closure Size58-400 mm
Closure TypeScrew Cap
Unit SizePack