Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Size |
|---|---|---|
| FIS02-896E | 500 mL | 55 mm |
| FIS02-896B | 60 mL | 28-415 |
| FIS02-896C | 125 mL | 38 mm |
| FKS11937934 | 125 mL | 38 mm |
| FIS02-896D | 250 mL | 45 mm |
| FKS11947934 | 250 mL | 45 mm |
| FIS02-896F | 1000 mL | 65 mm |
Completely disengage threads or remove cap before autoclaving.