Wide-Mouth Polypropylene Jars
Wide-Mouth Polypropylene Jars
Actual product may vary
Wide-Mouth Polypropylene Jars
Higher temperature resistance than LDPE or HDPE.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS02-891F950 mL120 mm
FIS02-891B60 mL53 mm
FIS02-891D250 mL89 mm
FIS02-891E500 mL89 mm
Catalog number FIS02-891F
Price (SGD)
312.00
CS
Add to cart
Capacity (Metric):
950 mL
Closure Size:
120 mm
Price (SGD)
312.00
CS
Add to cart
Higher temperature resistance than LDPE or HDPE.
  • Handle up to 135°C (275°F) for short periods and 100°C (212°F) continuously
  • Brittleness temperature of 0°C (32°F)
  • Rigid walls
  • Excellent chemical resistance
  • Can be sterilized by all normal methods, except dry heat and radiation
Completely disengage threads or remove cap before autoclaving.
Specifications
Capacity32 oz., 950 mL
Capacity (Metric)950 mL
Capacity (English)32 oz.
ColorNatural
DescriptionJar PP Natural 32oz
Diameter (English)4.7 in.
For Use With (Application)Storing Lab Specimen, Supplies or Powder
IncludesPolypropylene Linerless Screw Closure
MaterialPolypropylene
Max. Temperature (Metric)135°C
Max. Temperature (English)275°F
No. per Case24
No. per Pack6
QuantityCase, Pack
SterilityNon-sterile
Closure ColorWhite
ClarityNatural
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthWide
ShapeRound
Height (Metric)100 mm
Product TypeJar
Height (English)3.9 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size120 mm
Diameter (Metric)120 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS