Fisherbrand™ Large-Orifice Pipet Tips
Actual product may vary
Fisherbrand™ Large-Orifice Pipet Tips
Fisherbrand™ Large-Orifice Pipet Tips helps you pipet difficult liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatVolume (Metric)ColorLength (English)
FIS02-707-145Rack100 to 1000 μLNatural3.4 in.
FIS02-707-134Hinged Rack1 to 200 μLYellow2 in.
Catalog number FIS02-707-145
Price (SGD)
-
Format:
Rack
Volume (Metric):
100 to 1000 μL
Color:
Natural
Length (English):
3.4 in.
Genomic/Wide Orifice assists in pipetting small volumes of viscous materials.
  • Assist in pipetting small volumes of viscous materials
  • Help eliminate shearing of genomic DNA and fragile cells
Specifications
Volume (Metric)100 to 1000 μL
SterilitySterile
ColorNatural
Length (English)3.4 in.
Length (Metric)8.6cm
FormatRack
Tip StyleGenomic/Wide Orifice
PackagingLRS, 10 x 100
For Use With (Equipment)Sartorius Biohit Proline, Sartorius Biohit Proline Electronic, Transferpette, Costar Onepette, Eppendorf Research 3130, Eppendorf Reference 2000, Eppendorf Research 2100, Eppendorf, Thermo Scientific Finnpipette Digital
Unit SizePack