Search Fisher Scientific
| Catalog Number | Size |
|---|---|
| FIS02-682-271 | Medium |
| FIS02-682-270 | Small |
| 2008-ER | Large |
| FIS02-682-272 | Large |
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks. ART barrier tips provide 100% security against aerosols and liquid contamination. Front latch closes and locks, ensuring safe and secure storage throughout.
Features:
Certifications:
Recommended for: