Check Order Status
Nonsterile Threaded End Microcentrifuge Tubes without Caps
Actual product may vary
Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style.
Have Questions?
Catalog Number
Color
Capacity (Metric)
Max. RCF
Bottom Shape
FIS02-681-339
Natural
1.5 mL
30,000 x g
Conical
Catalog number
FIS02-681-339
Price (SGD)
72.00
Pack
Add to cart
Color:
Natural
Capacity (Metric):
1.5 mL
Max. RCF:
30,000 x g
Bottom Shape:
Conical
Price (SGD)
72.00
Pack
Add to cart
Choose skirted- or conical-bottom style.
Threaded to accept colored screw caps (
sold separately
)
Siliconized/low-retention conical-bottom tubes also available
Note:
For caps, see Fisherbrand™ Colored Screw Caps.
Specifications
Capacity (Metric)
1.5 mL
Color
Natural
Material
Plastic
Sterility
Non-sterile
Closure Type
Screw Cap
Bottom Shape
Conical
Max. RCF
30,000 x g