▸
Pipet Products
▸
Transfer Pipets
▸
Kimble™ Wintrobe Erythrocyte Sedimentation Rate (ESR) Tubes
Actual product may vary
DWK Life Sciences
Kimble™ Wintrobe Erythrocyte Sedimentation Rate (ESR) Tubes
Offers full calibration
Have Questions?
Catalog number
FIS02-681-1C
Price (SGD)
1,288.00
Pack
Add to cart
Price (SGD)
1,288.00
Pack
Add to cart
Cotton plugged
Full calibration in yellow
Fill with aspiration bulb No. 16
I.D. × L: 3 × 150mm (0.12 × 5.9 in.)
Specifications
Material
Soda Lime Glass
Quantity
100/Pk.
Length (Metric)
15 cm
Type
Wintrobe Pipet