Sedgwick-Rafter Cell Counting Chambers
Actual product may vary
Hausser Scientific
Sedgwick-Rafter Cell Counting Chambers
For In Vitro diagnostic use
Have Questions?
Catalog NumberLength (Metric)
FIS02-671-567 cm
Catalog number FIS02-671-56
Price (SGD)
1,134.00
Each
Length (Metric):
7 cm
Price (SGD)
1,134.00
Each
Specifications
Product TypeCounting Chamber
MaterialGlass
Length (English)2.7 in.
Length (Metric)7 cm
Thickness (Metric)0.05cm
Width (English)0.1 in.
Width (Metric)0.3cm
For Use With (Application)Microscopy of drinking Water, dust examination
For Use With (Equipment)Microscopy of drinking Water
IncludesCover Glass
Unit SizeEach