Bright-Line™ and Hy-Lite™ Counting Chambers
Actual product may vary
Hausser Scientific
Bright-Line™ and Hy-Lite™ Counting Chambers
For accurate white blood cell, red blood cell, and platelet counts
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial
FIS02-671-5Soda lime glass
FIS02-671-10Borosilicate Glass
Catalog number FIS02-671-5
Price (SGD)
810.00
Each
Material:
Soda lime glass
Price (SGD)
810.00
Each
For accurate white blood cell, red blood cell, and platelet counts
Difference in surface tension between cover glass and metal counting surface assures smooth capillary action for precise loading and more even cell distribution.
  • Reichert™ Bright-Line Counting Chambers are molded from quartz Borosilicate Glass
  • Hausser Bright-Line Counting Chambers are milled from soda lime glass
  • H-shaped moat forms two counting areas
  • Thin metal surface on each counting area has double Neubauer rulings with 400 small squares in central 1mm square
  • When viewed through microscope, rulings appear as white lines on dark background
  • Flat, polished cover glass covers moat region 0.1mm above ruled surface
  • V-shaped slashes at each loading side of counting chambers reduce overflow into moat
Specifications
Product TypeHemacytometer
Grid TypeReichert Bright Line
Capacity (Metric)0.1μL
MaterialSoda lime glass
QuantityEach
Depth (English)0.003 in.
Depth (Metric)0.01cm
For Use With (Application)Counting White Blood Cells, Red Blood Cells, and Platelets
IncludesTwo 0.4mm cover Glasses
Unit SizeEach