Hy-Lite™ Counting Chamber Set
Actual product may vary
Hausser Scientific
Hy-Lite™ Counting Chamber Set
Supplied with polyethylene carrying case
Have Questions?
Catalog NumberQuantity
FIS02-671-111 Ea.
Catalog number FIS02-671-11
Price (SGD)
816.00
Each
Quantity:
1 Ea.
Price (SGD)
816.00
Each
Supplied with polyethylene carrying case
Specifications
Product TypeHemacytometer Chamber Set
Grid TypeHausser Hy-Lite
Capacity (Metric)0.1μL
Quantity1 Ea.
Depth (English)0.003 in.
Depth (Metric)0.01cm
For Use With (Application)Consistent Cell Counting
IncludesCounting chamber, two cover Glasses, two pipets (one for red and one for white corpuscles) and pipet filler, Polyethylene carrying case
Unit SizeEach