Cell Counters
Actual product may vary
Bal Supply
Cell Counters
For automatic calculation including percentages.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStyle
FIS02-670-135 Counting units; 2 reset knob
FIS02-670-111 Counting unit; 1 reset knob
FIS02-670-122 Counting units; 1 reset knob
FIS02-670-148 Counting units; 2 reset knob
Catalog number FIS02-670-13
Price (SGD)
243.00
Each
Style:
5 Counting units; 2 reset knob
Price (SGD)
243.00
Each
For automatic calculation including percentages
  • Totaling unit rings a bell at 100 increments
  • Dual reset knobs
  • Last unit is the totaling unit, which records total number of strokes made on other keys
  • Removable tabs on top of counter provide area for identification of classification being counted
Specifications
Product TypeDifferential Cell Counter
Displayed Text0 to 999
Style5 Counting units; 2 reset knob
For Use With (Application)Cell Counting with Automatic Calculations
Unit SizeEach