DIFFCOUNT™ III Electronic Differential Tally Counters
DIFFCOUNT™ III Electronic Differential Tally Counters
Actual product may vary
DIFFCOUNT™ III Electronic Differential Tally Counters
Microprocessor-controlled differential cell counter designed specifically for hematology laboratory
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeVoltage
FIS02-669-48CDiff Cell Counter (12-channel)110V
FIS02-669-48BDiff Cell Counter (8-channel)110V
Catalog number FIS02-669-48C
Price (SGD)
2,932.00
Each
Add to cart
Product Type:
Diff Cell Counter (12-channel)
Voltage:
110V
Price (SGD)
2,932.00
Each
Add to cart
Microprocessor-controlled differential cell counter designed specifically for hematology laboratory
  • Displays all channels simultaneously with large digits on bright backlit LCD display
  • Hermetically sealed switches prevent count anomalies resulting from powdered gloves or other contaminants
  • Compact keyboard with “soft-touch” keys facilitates increased counting speed
  • Key assignment overlays available separately
  • Alarm actuates every 100 counts with optional 100-count lockout
  • Calculates percent and absolute WBC values with count correction
  • Operating temperature range: 0° to 49°C (32° to 120°F)
Model 10-308 feature eight counting keys, plus PERCENT, COUNT, and CLEAR keys. Counting and total displays have three digits
Model 10-312 and 10-312P feature 12 counting keys, plus PERCENT, COUNT, WBC, and CLEAR keys. Each counting channel displays two digits; total displays three digits. Model 10-312P includes a RS-232 printer port.
Specifications
Product TypeDiff Cell Counter (12-channel)
Display TypeLCD
Total Count12.0
Depth (English)9 in.
Depth (Metric)23cm
Height (English)2.25 in.
Height (Metric)6cm
Width (English)6 in.
Width (Metric)15cm
Frequency50/60Hz
Voltage110V
Wattage5w
For Use With (Application)Calculates percent or absolute WBC values
Unit SizeEach