Deluxe Beaker Tongs with Fiberglas™ - Covered Jaws
Deluxe Beaker Tongs with Fiberglas™ - Covered Jaws
Actual product may vary
Deluxe Beaker Tongs with Fiberglas™ - Covered Jaws
Fisherbrand Deluxe Beaker Tongs with Fiberglas™ - Covered Jaws safely and effectively handles beakers of volumes: 100-2000 mL.
Have Questions?
Catalog NumberLength (Metric)
FIS02-62233 cm
Catalog number FIS02-622
Price (SGD)
502.00
EA
Add to cart
Length (Metric):
33 cm
Price (SGD)
502.00
EA
Add to cart
  • Fisherbrand™ Deluxe Beaker Tongs with Fiberglas™ - Covered Jaws safely and effectively handles beakers of volumes: 100-2000 mL Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Designed for safe handling of beakers and vessels
  • Fiberglas™-covered jaws are 3 in. (7.6 cm) lower than handgrip
  • Hand is a safe 6 in. (15 cm) from beaker
  • Diecast aluminum hand grip for maximum comfort and control
  • Screw adjustment for maximum opening
  • Made from ASTM™ standard stainless-steel construction
  • Available without Fiberglass sleeve (Catalogue number: 02620)
Specifications
For Use With (Application)Safely handling 100 to 2000mL beakers
IncludesFiberglass covered jaws, Diecast Aluminum handgrip, screw adjustment
Length (English)13 in.
Length (English) Opening3 in.
Length (Metric) Opening7.6 cm
Length (Metric)33 cm
MaterialStainless Steel
TypeBeaker Tongs, Deluxe
Unit SizeEA