PFA Flasks with Screw Caps
Actual product may vary
PFA Flasks with Screw Caps
Calibrated 'To Contain' within Class A tolerances.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Tolerance
FIS02-617-156500 mL27 cm0.25 mL
FIS02-617-15225 mL11.5 cm0.04 mL
FIS02-617-155250 mL23.5 cm0.15 mL
Catalog number FIS02-617-156
Price (SGD)
-
Capacity (Metric):
500 mL
Height (Metric):
27 cm
Tolerance:
0.25 mL
Calibrated To Contain within Class A tolerances.
Suitable for trace element analysis, isotope separations, sample digestions, and the handling of sensitive biological solutions
  • Made from PFA copolymer
  • Translucent
  • Deliver outstanding chemical and solvent resistance over a temperature range of -260° to +270°C
  • Extremely low absorption and permeability levels associated with PFA
  • Graduation lines are highly legible, accurate, and permanent
  • Autoclavable at 250°F/121°C for 20 minutes
  • Can be sterilized using ethylene oxide gas, dry heat (160°C), beta or gamma irradiation (25kGy), or chemical disinfectant
Specifications
CalibrationCalibrated To Contain
Capacity (English)16.9 oz.
Closure MaterialPerfluoxy Copolymer
ColorHighly Translucent
DescriptionIndividually calibrated ring-marking
Capacity (Metric)500 mL
MaterialPFA
Height (English)10.6 in.
Height (Metric)27 cm
Product TypeVolumetric Flask
Temperature Range (English)-328°F to +500°F
Temperature Range (Metric)-200°C to +260°C
Tolerance0.25 mL
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
Mouth StyleThreaded
SterilityNon-sterile
Bottom ShapeRound
Unit SizeEach