PYREX™ Ribbed Watch Glass/Beaker Cover
Actual product may vary
PYREX™ Ribbed Watch Glass/Beaker Cover
Ribbed design permits reflux of digestate and escape of excess vapor, thus preventing vapor buildup.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)For Use With (Equipment)
FIS02-613C12.5 cmFor 1000 mL beaker
FIS02-613B10 cmFor 600 mL beaker
Catalog number FIS02-613C
Price (SGD)
612.00
Case
Diameter (Metric):
12.5 cm
For Use With (Equipment):
For 1000 mL beaker
Price (SGD)
612.00
Case
Designed for use as a beaker cover. Ribs are molded into the glass so that they can’t pop off as they do when cane is sealed to standard watch glasses. Permits reflux of digestate and escape of excess vapor. Eliminates risk of contamination from dropped hooks or props. Meets EPA requirements for methods 3005, 3010, and 3020.
Specifications
Certifications/ComplianceU.S. EPA
DisposableReusable
MaterialBorosilicate Glass
ShapeRound
TypeWatch Glass
For Use With (Application)Used as Beaker Covers
For Use With (Equipment)For 1000 mL beaker
Interior FinishRib
No. per Case12/Cs.
No. per Pack12/Pk.
Diameter (English)4.92 in.
Diameter (Metric)12.5 cm
Unit SizeCase