PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
Actual product may vary
PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
Molded for table stacking
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)For Use With (Equipment)
FIS02-612F15 cmFor 2000 mL beaker
FIS02-612E12.5 cmFor 1000 mL beaker
FIS02-612B7.5 cmFor 250 mL beaker
FIS02-612A6.5 cmFor 150 mL beaker
FIS02-612D10 cmFor 600 mL beaker
FIS02-612C9 cmFor 400 mL beaker
Catalog number FIS02-612F
Price (SGD)
347.00
PK
Add to cart
Diameter (Metric):
15 cm
For Use With (Equipment):
For 2000 mL beaker
Price (SGD)
347.00
PK
Add to cart
Molded for table stacking
These watch glasses possess high chemical durability, thus preventing contamination of solutions. They are molded to the same radius of curvature for convenient stacking. The heavy wall and ­uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.
Specifications
DisposableReusable
MaterialBorosilicate Glass
ShapeRound
TypeWatch Glass
For Use With (Application)Used as Beaker Covers
For Use With (Equipment)For 2000 mL beaker
Interior FinishPolished
No. per Case72/Cs.
No. per Pack12/Pk.
Diameter (English)5.9 in.
Diameter (Metric)15 cm
Unit SizePK