Search Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FIS02-591-9C | 3000 mL |
| 1223-1000 | 1000 mL |
| FIS02-591-9A | 1000 mL |
| 1223-3000 | 3000 mL |
Ensure resistance against acids, bases, aliphatic alcohols, aldehydes and esters with Thermo Scientific™ Nalgene™ PMP Graduated Beakers with Handles. These beakers have rectangular handles for easy carrying and pouring, and can withstand continuous use at high temperatures.