Polyurethane Ice Buckets
Polyurethane Ice Buckets
Actual product may vary
Polyurethane Ice Buckets
For use with liquid nitrogen, dry ice, water ice, and salt solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS02-591-45Blue
FIS02-591-46Red
FIS02-591-44Black
Catalog number FIS02-591-45
Price (SGD)
444.00
EA
Add to cart
Color:
Blue
Price (SGD)
444.00
EA
Add to cart
For use with liquid nitrogen, dry ice, water ice, and salt solutions.
  • Capacity: 1.2 gal. (4.5L)
  • Rigid polyurethane skinned-foam construction provides high durability and excellent insulating properties; buckets will not sweat or leak, and are unbreakable with normal use
  • Resistant to acetone and easy to clean—can be chemically disinfected
  • Withstand temperatures up to 95°C (203°F)
  • Octagonal design will not deform; stable when full
  • Molded handles enable safe handling and transport
  • Flush lid with recessed handle allows stacking for convenient storage
Specifications
Height (Metric)18.5 cm
Height (English)7.25 in.
Closure TypeLid
Capacity (English)1.2 gal.
For Use With (Application)Dry ice, water ice and salt solutions
DescriptionIce Bucket
MaterialPolyurethane
TypeIce Bucket
ColorBlue
Capacity (Metric)4.5 L
AutoclavableNot Autoclavable
Temperature Range95°C
Unit SizeEA