Tygon™ Sanitary Silicone Tubing Formula 3350
Actual product may vary
Saint-Gobain
Tygon™ Sanitary Silicone Tubing Formula 3350
Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (English) OuterThickness (English)
FIS02-587-1D0.094 in.0.16 in.0.031 in.
FIS02-587-1C0.063 in.3/16 in.0.0625 in.
Catalog number FIS02-587-1D
Price (SGD)
-
Diameter (English) Inner:
0.094 in.
Diameter (English) Outer:
0.16 in.
Thickness (English):
0.031 in.
Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications
Ultrasmooth inner bore reduces risk of particle entrapment, inhibits microscopic bacterial buildup during fluid transfer, and allows thorough cleaning and sterilization.
  • Tasteless, odorless
  • Nonpyrogenic, nontoxic, nonhemolytic
  • Low extractable content helps to maintain integrity and purity of transported fluids
  • Smooth inner surface provides low adsorption for improved fluid flow (hydrophobic surface also improves fluid flow)
  • Maximum recommended operating temperature 200°C (392°F)
  • Autoclavable; also can be radiation- and gas-sterilized
  • Durometer hardness: Shore A, 50
Not intended for use as an implant material.
Specifications
Certifications/ComplianceComplies with requirements of USP XXII Class VI criteria. Meets 3A Sanitary Standards, FDA 21 CFR Part 177.2600 criteria and NSF 51 Standard.
AutoclavableAutoclavable
Diameter (English) Inner0.094 in.
Diameter (Metric) Inner2.4 mm
MaterialTygon
Diameter (English) Outer0.16 in.
Diameter (Metric) Outer4 mm
Overall Length (English)50 ft.
Thickness (English)0.031 in.
Thickness (Metric)0.8 mm
Unit SizePack