Specifications
Capacity (English)8.45 oz.
Capacity (Metric)250 mL
MaterialPTFE
For Use With (Application)Ideal for Digestion
Height (English)3.63 in.
Height (Metric)9.23 cm
IncludesSpout
Temperature (English) Max. Operating550°F
Temperature (Metric) Max. Operating288°C
TypeBeaker
StyleGriffin
Unit SizeEach