Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD
Actual product may vary
Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD
Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD
DWK Life Sciences
Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD
Designed from ASTM Specification E969, Class A requirements. Kimble™ Reusable Class A Volumetric Serialized and Certified Pipets, TD are ideal for measuring accurate volumes of liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ToleranceLength (Metric)
FIS02-555-365100 mL±0.08 mL55 cm
FIS02-555-35920 mL±0.03 mL50.5 cm
FIS02-555-36025 mL±0.03 mL50.5 cm
FIS02-555-36350 mL±0.05 mL54.5 cm
Catalog number FIS02-555-365
Price (SGD)
605.00
Case
Volume (Metric):
100 mL
Tolerance:
±0.08 mL
Length (Metric):
55 cm
Price (SGD)
605.00
Case
Designed from ASTM Specification E969, Class A requirements. Kimble™ KIMAX™ Reusable Class A Volumetric Serialized Pipets, TD are ideal for measuring accurate volumes of liquids.
  • Includes individual Certificates of Calibration
  • Sizes 1mL and larger are marked with large numerals on the bulb, indicating capacity
  • Color-coded (ASTM E1273) for ease in sorting and selecting the correct pipet
  • With a legend of permanent brown stain
  • Permanently marked individual serial number
    • Autoclavable 
    • 75 mL size designed from ASTM E542 
    • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
AutoclavableAutoclavable
CalibrationTo Deliver
Certifications/ComplianceCertificate of graduation accuracy, ASTM E969, ASTM E542
ClassA
MaterialBorosilicate Glass
Tolerance±0.08 mL
ColorYellow
Volume (Metric)100 mL
Length (Metric)55 cm
TypeSerialized Volumetric Pipet