Automatic Glass Titrating Burets
Automatic Glass Titrating Burets
Actual product may vary
Dynalon
Automatic Glass Titrating Burets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-550-10325 mL
FIS02-550-10450 mL
Catalog number FIS02-550-103
Price (SGD)
524.00
EA
Add to cart
Capacity (Metric):
25 mL
Price (SGD)
524.00
EA
Add to cart
Simple and accurate to use. Squeeze the flexible LDPE bottle until the titrate solution flows through the tubing into the glass buret to just above the “0” graduation mark. Product is designed to self zero and does not overflow.
Specifications
Capacity (Metric)25 mL
Height (English)24.1 in.
For Use With (Equipment)1L LDPE bottle
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)Electroplating Industry, Field Analyses and Numerous Other Applications, Universal Aid in the Laboratory, Water Treatment Plants
Height (Metric)61 cm
IncludesStand, Blue Ring Graduations, Schellbach Stripe
ScaleBlue
Stopcock MaterialLPDE
Tolerance±0.1 mL
TypePellet Pattern Automatic Buret
ClassClass B
Certifications/ComplianceDIN ISO 385
Unit SizeEA