Multipurpose Sample Cups
Actual product may vary
Multipurpose Sample Cups
Heavy-duty polystyrene to prevent cracking or tipping
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Capacity (English)
FIS02-544-192 mL0.06 oz.
FIS02-544-44 mL0.13 oz.
Catalog number FIS02-544-19
Price (SGD)
-
Capacity (Metric):
2 mL
Capacity (English):
0.06 oz.
Heavy-duty polystyrene to prevent cracking or tipping
  • Graduated in 0.5mL increments
  • Side-gated molding process eliminates pinholes in bottoms of cups
Specifications
Capacity (Metric)2 mL
Product TypeSample Cup
Capacity (English)0.06 oz.
MaterialPolystyrene
For Use With (Application)To prevent cracking or tipping
Capacity2 mL
Unit SizePack