▸
Stoppers and Closures
▸
Bottle Caps
▸
Kimble™ Blue Polypropylene Cap
Actual product may vary
DWK Life Sciences
Kimble™ Blue Polypropylene Cap
Durable, screw-thread caps provide secure closure
Have Questions?
Catalog Number
Thread Size (Metric)
FIS02-542-1
GL 45
Catalog number
FIS02-542-1
Price (SGD)
159.00
Case
Add to cart
Thread Size (Metric):
GL 45
Price (SGD)
159.00
Case
Add to cart
Durable, screw-thread caps provide secure closure
Solid top polypropylene GL 45 caps with internal molded seal rings
Color: Blue
Made of polypropylene
Specifications
Color
Blue
Material
Polypropylene
No. per Case
10 Cs.
Thread Size (Metric)
GL 45
Neck Finish
43 mm
Product Type
Screw Cap
Autoclavable
Autoclavable