Disposable Tissue Grinders
Actual product may vary
Disposable Tissue Grinders
Disposable Tissue Grinders
Disposable Tissue Grinders
Closed tissue grinder system minimizes sample contact by aerosols and sample overflow
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityProduct TypeAutoclavable
FIS02-542-071.69 oz., 50mLTissue GrinderAutoclavable
FIS02-542-111.69 oz., 50mLUltra Tissue GrinderAutoclavable
FIS02-542-090.5 oz., 15mLTissue GrinderNot Autoclavable
Catalog number FIS02-542-07
Price (SGD)
-
Capacity:
1.69 oz., 50mL
Product Type:
Tissue Grinder
Autoclavable:
Autoclavable
  • Sterile and pre-assembled
  • Closed system prevents risk of contact with sample
  • Easy-to-open Tyvek™ packaging
  • Extra cap and patient label provided
Standard Tissue Grinders
  • Feature a molded-in abrasive surface; no sediment to obscure sample
  • Available in 15, 35 and 50mL sizes
Ultra Tissue Grinders
  • Silica cast tip provides enhanced grinding for difficult samples
  • Available in 15 and 50mL sizes
Specifications
Capacity (English)1.69 oz.
For Use With (Application)Grinding small tissue samples
AutoclavableAutoclavable
DisposableSingle-use
IncludesPolypropylene tube, polystyrene pestle and tip, HDPE cap and extra cap
Capacity (Metric)50 mL
Capacity1.69 oz., 50mL
SterilitySterile
Product TypeTissue Grinder
Unit SizeCase