PYREX™ Low Form Griffin Beakers
Actual product may vary
PYREX™ Low Form Griffin Beakers
Offers optimum balance between thermal shock resistance and mechanical strength
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-540K250 mL
Catalog number FIS02-540K
Price (SGD)
177.00
Pack
Capacity (Metric):
250 mL
Price (SGD)
177.00
Pack
With pouring spout and extra-large marking spot
Corning™ PYREX™ Low Form Griffin Beakers offer optimum balance between thermal shock resistance and mechanical strength.
With pouring spout and extra-large marking spot.
Beakers, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10 mL size is not graduated.
Specifications
Capacity (English)8.45 oz.
Diameter (English) Outer2.67 in.
Diameter (Metric) Outer6.8 cm
Capacity (Metric)250 mL
MaterialBorosilicate Glass
For Use With (Application)For Optimum Balance Between Thermal Shock Resistance and Mechanical Strength
Graduation Range25 to 200 mL
Height (English)3.54 in.
Height (Metric)9 cm
IncludesSpout
Increments25 mL
Label TypeWhite Marking
Quantity12 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleDual White Metric Scale
StyleGriffin
Unit SizePack