Nalgene™ Coex Polyethylene Film Tank Liners
Nalgene™ Coex Polyethylene Film Tank Liners
Actual product may vary
Thermo Scientific
Nalgene™ Coex Polyethylene Film Tank Liners
Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)
FIS02-540-61315 gal.
FIS02-540-6117.5 gal.
FIS02-540-61430 gal.
FIS02-540-61680 gal.
Catalog number FIS02-540-613
Price (SGD)
2,907.00
CS
Add to cart
Capacity (English):
15 gal.
Price (SGD)
2,907.00
CS
Add to cart
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.
  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Warranty:
90 days
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeTank Liner
DescriptionGamma Irradiated: 15 gal. (57L)
MaterialPolyethylene
Capacity (English)15 gal.
Capacity (Metric)57 L
For Use With (Equipment)Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case10
No. per Pack10
Unit SizeCS