Individual Analytical Precision Weights, Class 1 with NVLAP Certificate
Actual product may vary
Troemner
Individual Analytical Precision Weights, Class 1 with NVLAP Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMass
FIS02-319-1BC50 g
FIS02-214-1JW200 g
FIS02-319-1AC100 g
Catalog number FIS02-319-1BC
Price (SGD)
596.00
Each
Mass:
50 g
Price (SGD)
596.00
Each
Designed so the specifications on surface finish, markings, construction, material density and magnetic properties get more restrictive as the weight tolerances get smaller in order to enable the weight to maintain its tolerance over time
  • Used in extremely stable environments
  • 1mg to 500mg are 1 piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are 2 piece construction with handling knob
  • Surfaces are polished to a perfect, mirror-like finish
Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.1mg to 0.01mg
Specifications
Product TypeTwo-Piece Metric Analytical Weight
Certifications/ComplianceNVLAP, ISO9001
ClassificationASTM Class 1
MaterialStainless Steel
Weight (Metric)50 g
Density7.95g/cm3 at 20°C
Mass50 g
ShapeCylindrical with Handling Knob
IncludesPolycarbonate case
QuantityEach
Packaging Quantity1
Unit SizeEach