▸
Clamps and Supports
▸
Clamps
▸
Stainless Steel Double Buret Clamp and Support Stand with Rod
Actual product may vary
Troemner
Stainless Steel Double Buret Clamp and Support Stand with Rod
Double buret clamp gently but firmly holds two burets at once.
Have Questions?
Catalog Number
ForUseWithEquipment
FIS02-300-254
Catalog number
FIS02-300-254
Price (SGD)
1,665.00
Each
Add to cart
Price (SGD)
1,665.00
Each
Add to cart
Double buret clamp gently but firmly holds two burets at once.
Double Buret Clamp holds any size buret from micro to 100mL capacity
Compress the scissor-like mechanism, insert buret and gently release to grip
Numbers and graduation on buret remain easy to read
For height adjustments, recompress mechanism and slide buret up or down and gently release
Unit attaches to optional standard support rod with built-reinforced hook connector
Adjusting nut faces forward for easy use
Stainless steel rod attaches to optional porcelain base
Clamp dimensions: 173 x 108mm (6.8 x 4.25 in.)
Rod diameter x length: 13 x 578mm (0.51 x 22.76 in.)
Base dimensions: 178 x 330 x 25mm (7 x 13 x 1 in.)
Specifications
Quantity
1 Ea.
Type
Double Burette Clamp
Material
Stainless Steel (Electro-polished)
Holds
Beakers, 50 to 2000 mL
Includes
Stainless Steel Support Rod, Porcelain Base
Diameter (English)
1/4 in.
Length (English)
9.5 in.
Size (Metric) Grip
50 to 2000 mL