Nickel-plated zinc Double Buret Clamp holds any size buret from micro to 100mL capacity. Simply compress the scissor-like mechanism, insert buret and gently release to grip. Clamp only; rod and support stand available separately.
Have Questions?
Catalog Number
For Use With (Equipment)
FIS02-300-213
Labjaws
Catalog number FIS02-300-213
Price (SGD)
295.00
Each
For Use With (Equipment):
Labjaws
Price (SGD)
295.00
Each
Numbers and graduation on buret remain easy to read
For height adjustments, recompress mechanism and slide buret up or down and gently release
Double Buret Clamp unit attaches to optional standard support rod with built-reinforced hook connector
Adjusting nut faces forward for easy use
Stainless steel or aluminum support rod attaches to optional porcelain base