Two-Prong Single Adjust Multi-Purpose Clamp
Actual product may vary
Troemner
Two-Prong Single Adjust Multi-Purpose Clamp
Designed to securely hold laboratory glassware and apparatus. Extension armattaches clamp head securely and offers easy positioning in the deepest fumehoods.
Have Questions?
Catalog NumberSizeSize (Metric) GripSize (English) GripLength (Metric)
FIS02-300-205Large0 to 92 mm0 to 3.62 in.229 mm
Catalog number FIS02-300-205
Price (SGD)
180.00
Each
Size:
Large
Size (Metric) Grip:
0 to 92 mm
Size (English) Grip:
0 to 3.62 in.
Length (Metric):
229 mm
Price (SGD)
180.00
Each
  • Constructed with round extension arms, which allow the clamps to be rotated 360°
  • Extension arms also allow placement of apparatus at various distances from lab-frames without compromising the integrity of experiments
  • Nickel-plated zinc offers a clamp with a high tensile strength at an economical price
  • Supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers
Specifications
TypeMulti-purpose Clamp
MaterialNickel-plated Zinc
IncludesNon-slip vinyl sleeves, and for temperatures above 100°C, fiberglass covers
For Use With (Application)Securely hold laboratory glassware and apparatus
Diameter (English)0.43 in.
Diameter (Metric)11 mm
Length (English)9.02 in.
Length (English) Arm5 in.
Length (Metric)229 mm
Length (Metric) Arm127 mm
SizeLarge
Size (Metric) Grip0 to 92 mm
Size (English) Grip0 to 3.62 in.
Jaw Type2-Prong Single Adjust
Unit SizeEach