Ultra Flex Clamping Systems
Actual product may vary
Ultra Flex Clamping Systems
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeIncludes
FIS02-217-00712 in. (Arm)Base Plate
FIS02-217-00913 in. (Arm)Lab Frame Connector
Catalog number FIS02-217-007
Price (SGD)
780.00
Each
Size:
12 in. (Arm)
Includes:
Base Plate
Price (SGD)
780.00
Each
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
  • Ultra Flex comes in three different systems: base plate, lab-frame connector, and bench clamp
  • Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305mm (12 in.) or 457mm (18 in.) length, and utility wrench
  • 2-prong and 3-prong clamp heads are also supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers
Specifications
Quantity1 Ea.
TypeUltra Flex Clamp System
MaterialSteel (Base Plate)
IncludesBase Plate
For Use With (Equipment)Ultra flex clamping system components
DescriptionUltra Flex 12 with Base Plate
Length (English) Arm12 in.
Length (Metric) Arm30.48 cm
Size12 in. (Arm)
Size (Metric) Grip7.49, 6.9, 1.29 cm
Size (English) Grip2.95, 2.72, 0.51 in.
Jaw Type3-Prong
Unit SizeEach