Search Fisher Scientific
| Catalog Number | Diameter (Metric) Inside |
|---|---|
| FIS02-216-373 | 15 cm |
| FIS02-216-372 | 11.4 cm |
Hand-operated inoculating turntables
Talboys hand-operated inoculating turntables produce almost concentric circles of bacterial colonies that are evenly distributed across Petri dishes. Cast iron turntables are coated with gray baked acrylic enamel.
The large turntable accommodates 100mm or 150mm Petri dishes. It has a height of only 32mm (1.26 in.), ideal for steadying a forearm on the work surface while plating. The large turntable contains two circular sections. The top section has a raised gripper lip as well as a recessed center disk covered with a non-skid rubber lining.