Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Clamps
▸
Talboys™ Labjaws™ Multi-Purpose Column Clamps
Actual product may vary
Troemner
Talboys™ Labjaws™ Multi-Purpose Column Clamps
ideal for holding large cylindrical glassware and similar objects
Have Questions?
Change view
Catalog Number
Size
Size (English) Grip
Length (English)
FIS02-216-301
Medium
4.49 in.
12.4 in.
FIS02-216-302
Large
5.51 in.
13.31 in.
2 Options
Catalog number
FIS02-216-301
Price (SGD)
342.00
Each
Add to cart
Size:
Medium
Size (English) Grip:
4.49 in.
Length (English):
12.4 in.
Price (SGD)
342.00
Each
Add to cart
Sturdy, easily adjusted, multi-purpose clamps are ideal for holding large cylindrical glassware and similar objects
A large, flat thumbscrew/worm-drive permits maximum tightening or removing in minimum time
Constructed of corrosion-resistant stainless steel
Ideal for chromatography columns
Specifications
Type
Chain Clamp
Material
Stainless Steel
Length (English)
12.4 in.
Length (English) Arm
8.31 in.
Length (Metric)
31.49 cm
Length (Metric) Arm
21.1 cm
Size
Medium
Size (Metric) Grip
11.4 cm
Size (English) Grip
4.49 in.