Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Clamps
▸
Talboys™ Labjaws™ Double Buret Clamp with Stand
Actual product may vary
Troemner
Talboys™ Labjaws™ Double Buret Clamp with Stand
Securely grips burets from micro to 100mL capacities
Have Questions?
Catalog Number
For Use With (Equipment)
FIS02-216-299
Labjaws
Catalog number
FIS02-216-299
Price (SGD)
1,164.00
Each
Add to cart
For Use With (Equipment):
Labjaws
Price (SGD)
1,164.00
Each
Add to cart
Double buret clamp grasps burets from micro to 100mL capacities
Simple construction allows users to compress scissor-like mechanism, insert buret and gently release to grip
For height adjustments, recompress mechanism and slide buret up or down and gently release
Clamp unit attaches to standard support rod with built-in reinforced hook connector
Adjusting nut faces forward for easy height adjustment
Support stand comes with white porcelain base and a 0.51 x 22.76 in. (13 x 578mm) aluminum rod
Porcelain base measures 7 x 13 x 1 in. (17.8 x 33 x 2.5cm)
Parts are available individually or as a package
Specifications
Type
Double Burette Clamp and Stand
Material
Nickel, Zinc, Aluminum, Porcelain
Includes
Rod
For Use With (Equipment)
Labjaws