Talboys™ Aluminum Lab Lifts
Actual product may vary
Troemner
Talboys™ Aluminum Lab Lifts
Aluminum Lab-Lifts provide stable height adjustment for various items in the lab such as flasks, baths, and small equipment. Lab-Lifts accept optional Support Rod Kit, which mounts to the upper deck.
Have Questions?
Catalog NumberCapacity (English)
FIS02-214-211132 lb.
Catalog number FIS02-214-211
Price (SGD)
986.00
Dozen
Capacity (English):
132 lb.
Price (SGD)
986.00
Dozen
  • Exceptional stability and durability
  • Aluminum construction
  • Three convenient sizes
Top and bottom decks are constructed of anodized aluminum. Internal supports and drive screws are constructed of stainless steel. Oversized sure-grip adjustment knobs provide smooth and accurate height adjustment.
Specifications
MaterialAnodized Aluminum (Top and Bottom Decks), Stainless Steel (Internal Supports and Drive Screws)
Height (Metric)76 to 248 mm
Height (English)3 to 9.75 in.
Length (English) Base6 in.
Length (Metric) Base152 mm
Width (English) Base6 in.
Width (Metric) Base152 mm
Capacity (English)132 lb.
Product TypeLab-Lift
Unit SizeDozen