Weighing Canoes
Actual product may vary
Dyn-A-Med
Weighing Canoes
Resist dilute acids, aqueous solutions, alcohols and bases
Have Questions?
Catalog NumberLength (Metric)Width (Metric)
FIS02-202F7.3 cm12.1 cm
Catalog number FIS02-202F
Price (SGD)
150.00
Pack
Length (Metric):
7.3 cm
Width (Metric):
12.1 cm
Price (SGD)
150.00
Pack
  • Anti-static
  • Polystyrene
  • Contoured sides
Specifications
MaterialPolystyrene
TypeWeighing Dish Canoe
For Use With (Application)Ideal for weighing static-affected materials, either liquid or dry
Dimensions (L x W x H)2.88 x 4.75 x 1.25 in. (7.3 x 12.1 x 3.18 cm)
Length (Metric)7.3 cm
Width (Metric)12.1 cm
Height (Metric)3.18 cm
Unit SizePack