▸
Salts and Inorganics
▸
Inorganic Salts
▸
Transition Metal Salts
▸
Ferric Ammonium Sulfate, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Element Search
Structure Search
Ferric Ammonium Sulfate, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Have Questions?
Catalog Number
Quantity
Packaging
FIS02-002-860
500 g
Poly Bottle
Catalog number
FIS02-002-860
Price (SGD)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ferric Ammonium Sulfate
Name Note
12-Hydrate
CAS
7783-83-7
Physical Form
Crystals
Quantity
500 g
View more