Search Thermo Fisher Scientific
| Catalog Number | Color | Dimensions (L x W) | Thickness |
|---|---|---|---|
| FIS01829C | Red with Black Print | 23 x 19 in. (58 x 48 cm) | 1.50 mil |
| FIS01-814-1 | Natural | 24 x 12 in. (61 x 30 cm) | 1.50 mil |
| FIS01-814-3 | Natural | 24 x 36 in. (61 x 91 cm) | 1.50 mil |
| FIS01-827A | Clear with Red Print | 12 x 8 in. (30 x 20 cm) | 1.50 mil |
| FIS01-827D | Clear with Red Print | 36 x 24 in. (91 x 61 cm) | 1.50 mil |
| FIS01-827B | Clear with Red Print | 24 x 12 in. (61 x 30 cm) | 1.50 mil |
| FIS01-827C | Clear with Red Print | 30 x 24 in. (76 x 61 cm) | 1.50 mil |
| FIS01-829A | Red with Black Print | 12 x 8 in. (30 x 20 cm) | 1.50 mil |
| FIS01-829B | Red with Black Print | 19 x 14 in. (48 x 36 cm) | 1.50 mil |
| FIS01-829C | Red with Black Print | 23 x 19 in. (58 x 48 cm) | 1.50 mil |
| FIS01-829D | Red with Black Print | 25 x 35 in. (64 x 89 cm) | 1.50 mil |
| FIS01-829F | Red with Black Print | 30 x 33 in. (76 x 84 cm) | 1.50 mil |
| FIS01-831B | Red with Black Print | 48 x 38 in. (122 x 97 cm) | 4 mil |
| FIS01-829E | Red with Black Print | 48 x 38 in. (122 x 97 cm) | 1.50 mil |