InLab™ Pure pH Electrodes
Actual product may vary
InLab™ Pure pH Electrodes
InLab™ Pure pH Electrodes
InLab™ Pure pH Electrodes
InLab™ Pure pH Electrodes
InLab™ Pure pH Electrodes
InLab™ Pure pH Electrodes
InLab™ Pure pH Electrodes
Mettler Toledo
InLab™ Pure pH Electrodes
A glass pH electrode designed for ion-weak-samples such as ultrapure water and deionized water.
Have Questions?
Catalog NumberConnector Types
FIS01-917-146Multi-Pin
Catalog number FIS01-917-146
Price (SGD)
3,208.00
Each
Connector Types:
Multi-Pin
Price (SGD)
3,208.00
Each
  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
  • Pure water
  • Ultrapure water
  • Deionized water
  • Waste water
  • Highly saline solution, sea water
  • Low-ion strength
Accessories & Care
  • Immovable glass sleeve junction
  • LoT membrane glass
  • InLab Pure: refillable FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Pure Pro-ISM: refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
DescriptionpH electrode InLab Pure Pro-ISM
Connector TypesMulti-Pin
JunctionImmovable Glass Sleeve
MaterialGlass
pH Range1 to 11
Temperature (Metric) Operating0°C to 80°C
TypeCombination Electrode
Filling Solution Type3 mol/L KCl Reference, 1mol/L KCl Bridge
For Use With (Application)Measuring samples at low temperatures or with low ion concentrations
For Use With (Equipment)Multi-Pin to BNC/RCA (30281896)
IncludesISM Temperature Probe
Internal ReferenceARGENTHAL with Ag+ trap
MembraneLoT Glass
ModelInLab™ Pure Pro-ISM
ProbeNTC 30 kω
Storage Requirements3mol/L KCl
Temperature (English) Operating32°F to 176°F
Temperature (Metric) Range0°C to 80°C
Length (English)6.7 in.
Length (Metric)170 mm
Diameter (English)0.47 in.
Diameter (Metric) Body12 mm
Unit SizeEach