Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™
Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™
Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™
Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™
METTLER TOLEDO™ solutions for electrode calibration and care.
Have Questions?
Catalog NumberQuantity
FIS01-917-117500 mL
Catalog number FIS01-917-117
Price (SGD)
113.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
113.00
Each
Specifications
Chemical Name or MaterialBuffer Solution, pH 4.00
CAS877-24-7
Melting Point0°C
Boiling Point100°C
ColorRed
View more
  • Quality solutions for accurate results
  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards