EasyPlus™ Titration Sensors
Actual product may vary
EasyPlus™ Titration Sensors
EasyPlus™ Titration Sensors
Mettler Toledo
EasyPlus™ Titration Sensors
Specially developed for use with the EasyPlus Titrators.
Have Questions?
Catalog NumberTypeReference Electrolyte
FIS01-912-526EG11-BNC Sensor pH aqueous 3mol/L KCl
Catalog number FIS01-912-526
Price (SGD)
855.00
Each
Type:
EG11-BNC Sensor pH aqueous
Reference Electrolyte:
3mol/L KCl
Price (SGD)
855.00
Each
  • Used for routine acid/base application in aqueous media and non-aqueous solutions
  • Delivers quality and reliable results
  • Made of glass
pH aqueous Sensor
  • Ideal for acid/base titrations in aqueous media
  • Determines acidity in juice, milk and wine
  • Simultaneously measures m- and p-value in water
pH non-aqueous Sensor
  • Used in acid/base titrations in non-aqueous media such as free fatty acid content of edible oils
Specifications
Connector TypesBNC Fixed Cable
TypeEG11-BNC Sensor pH aqueous
Internal ReferenceConductor
JunctionCeramic
MaterialGlass
Reference Electrolyte3mol/L KCl
Temperature (English) Range0°F to 140°F
Temperature (Metric) Range0°C to 60°C
Unit SizeEach